trans-2-Aminocyclohexanol hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning

20698-0050 20698-0250
TS20698-0050EA 93 USD
TS20698-0050 TS20698-0250
trans-2-Aminocyclohexanol hydrochloride 99%
trans-2-Aminocyclohexanol hydrochloride
Formula: C₆H₁₄ClNO
Melting Pt: 173…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00001490
CAS Number: 5456-63-3

Order Now

Specification Test Results

Appearance White to beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 173 to 177 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR