Naringin 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (S)-7-(((2S,3R,4S,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-5-hydroxy-2-(4-hydroxyphenyl)chroman-4-one, 4',5,7-Trihydroxyflavanone 7-rhamnoglucoside, Naringenin 7-rhamnoglucoside

20691-0250 20691-1000
TS20691-0250EA 55 USD
TS20691-0250 TS20691-1000
Naringin 97%
Naringin
Formula: C₂₇H₃₂O₁₄
Melting Pt: 166 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00148888
CAS Number: 10236-47-2

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Specific optical rotation -77° to -93° (20°C, 589 nm) (c=1, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR