Dicoumarol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,3'-Methylenebis(4-hydroxycoumarin), Bishydroxycoumarin, Dicoumarin
20412-0050
20412-0250
TS20412-0050EA
43
USD
TS20412-0050
TS20412-0250
Dicoumarol 99%
Dicoumarol
Formula:
C₁₉H₁₂O₆ MW: 336.30 g/mol Melting Pt: 290…292 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006857 CAS Number: 66-76-2 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with TBAH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...