Tetraphenylphosphonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

20403-0500 20403-0100
TS20403-0500EA 294 USD
TS20403-0500 TS20403-0100
Tetraphenylphosphonium chloride 98%
Tetraphenylphosphonium chloride
Formula: C₂₄H₂₀ClP
Melting Pt: 275…280 °C
MDL Number: MFCD00011916
CAS Number: 2001-45-8
EINECS: 217-890-3

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 275 to 280 °C
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR