2-Bromo-2-butene cis- and trans-mixture 98%

Supplier: Thermo Scientific Chemicals
Danger

20151-0250 20151-1000
TS20151-0250EA 83 USD
TS20151-0250 TS20151-1000
2-Bromo-2-butene cis- and trans-mixture 98%
2-Bromo-2-butene
Formula: C₄H₇Br
MW: 135 g/mol
Boiling Pt: 82…90 °C (740 mmHg)
Density: 1.32 g/cm³ (20 °C)
Flash Pt: <1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000141
CAS Number: 13294-71-8
EINECS: 236-316-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4620 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR