4-Iodo-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

20037-0050 20037-0250
TS20037-0050EA 104 USD
TS20037-0050 TS20037-0250
4-Iodo-1H-pyrazole 98%
4-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005244
CAS Number: 3469-69-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 107 to 112 °C
HPLC ≥97.5 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR