Bromoacetyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

19799-0250 19799-1000
TS19799-0250EA 75 USD
TS19799-0250 TS19799-1000
Bromoacetyl chloride 95%
Bromoacetyl chloride
Formula: BrCH₂COOCl
MW: 157.39 g/mol
Boiling Pt: 127…128 °C (760 mmHg)
Density: 1.9 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00000724
CAS Number: 22118-09-8
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4970 (20°C, 589 nm)
Titration Argentometric 94.0 to 106.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR