2-Bromo-6-methoxynaphthalene 98%

Supplier: Thermo Scientific Chemicals
Warning

19056-0050 19056-0250
TS19056-0050EA 40 USD
TS19056-0050 TS19056-0250
2-Bromo-6-methoxynaphthalene 98%
2-Bromo-6-methoxynaphthalene
Formula: C₁₁H₉BrO
MW: 237.1 g/mol
Melting Pt: 105…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00004062
CAS Number: 5111-65-9
EINECS: 225-837-0

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 105 to 111 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR