3-(3,4,5-Trimethoxyphenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4,5-Trimethoxyhydrocinnamic acid, 3-(3,4,5-Trimethoxyphenyl)propanoic acid

18916-0050 18916-0250
TS18916-0050EA 47 USD
TS18916-0050 TS18916-0250
3-(3,4,5-Trimethoxyphenyl)propionic acid 98%
3-(3,4,5-Trimethoxyphenyl)propionic acid
Formula: C₁₂H₁₆O₅
MW: 240.26 g/mol
Melting Pt: 100…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00002775
CAS Number: 25173-72-2
EINECS: 246-706-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 105 °C
HPLC ≥97.5 %
Dnase,Rnase,Protease activity none detected
Phosphate (PO4) ≤2 ppm
Sulfate (SO4) ≤20 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR