3-Fluoropyridine 99%

Supplier: Thermo Scientific Chemicals
Danger

18830-0010 18830-0050
TS18830-0010EA 22 USD
TS18830-0010 TS18830-0050
3-Fluoropyridine 99%
3-Fluoropyridine
Formula: C₅H₄FN
MW: 97.09 g/mol
Boiling Pt: 107 °C (1013 hPa)
Density: 1.13 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00006374
CAS Number: 372-47-4
EINECS: 206-755-4
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR