Thiomorpholine 97%

Supplier: Thermo Scientific Chemicals
Danger

18797-0050 18797-0250
TS18797-0050EA 52 USD
TS18797-0050 TS18797-0250
Thiomorpholine 97%
Thiomorpholine
Formula: C₄H₉NS
MW: 103.19 g/mol
Density: 1.088 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00005974
CAS Number: 123-90-0
UN: 3267
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5390 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR