4-Phenylcyclohexanone 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane

18794-0100 18794-0500
TS18794-0100EA 55 USD
TS18794-0100 TS18794-0500
4-Phenylcyclohexanone 98+%
4-Phenylcyclohexanone
Formula: C₁₂H₁₄O
MW: 174.24 g/mol
Melting Pt: 77…80 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00001641
CAS Number: 4894-75-1
EINECS: 225-517-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 76 to 80 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR