Thiophene-2-carbonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

18777-0250 18777-1000
TS18777-0250EA 38 USD
TS18777-0250 TS18777-1000
Thiophene-2-carbonyl chloride 98%
Thiophene-2-carbonyl chloride
Formula: C₅H₃ClOS
MW: 146.6 g/mol
Density: 1.37 g/cm³
Flash Pt: 90 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005428
CAS Number: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to green Liquid
Infrared spectrum Conforms
Refractive index 1.5890 to 1.5910 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR