2,5-Dihydroxy-p-benzoquinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
18746-0250
18746-1000
TS18746-0250EA
90
USD
TS18746-0250
TS18746-1000
2,5-Dihydroxy-p-benzoquinone 98%
2,5-Dihydroxy-p-benzoquinone
Formula:
C₆H₄O₄ Melting Pt: 220 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00001598 CAS Number: 615-94-1 EINECS: 210-454-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Ochre to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Titration with NaOH | 9.3 to 9.7 % (fumaric acid dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...