1-Bromotetradecane 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Myristyl bromide
18738-1000
18738-5000
TS18738-1000EA
53
USD
TS18738-1000
TS18738-5000
1-Bromotetradecane 98%
1-Bromotetradecane
Formula:
BrH₂C(CH₂)₁₂CH₃ Boiling Pt: 175…178 °C (20 mmHg) Melting Pt: 5…6 °C Density: 0.93 g/cm³ Flash Pt: 149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000228 CAS Number: 112-71-0 EINECS: 203-999-3 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4620 (20°C, 589 nm) |
GC | ≥97.5 % |
Ethanol | 95 to 97 % v/v (calculated) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...