2,2-Dibromopropane 98%

Supplier: Thermo Scientific Chemicals
Warning

18606-0050 18606-0500
TS18606-0050EA 91 USD
TS18606-0050 TS18606-0500
2,2-Dibromopropane 98%
2,2-Dibromopropane
Formula: H₃CBr₂CCH₃
MW: 201.89 g/mol
Boiling Pt: 114 °C (740 mmHg)
Melting Pt: –20 °C
Density: 1.78 g/cm³
Flash Pt: >110 °C
MDL Number: MFCD00000119
CAS Number: 594-16-1
EINECS: 209-828-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4990 (20°C, 589 nm)
GC ≥97.5 %
Residue after ignition ≤0.1 %
Specific optical rotation -66° to -69° (20°C, 589 nm) (1 % in EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR