Ethyl succinyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 4-chloro-4-oxobutanoate
18409-0250
18409-1000
TS18409-0250EA
71
USD
TS18409-0250
TS18409-1000
Ethyl succinyl chloride 95%
Ethyl succinyl chloride
Formula:
C₆H₉ClO₃ MW: 164.59 g/mol Boiling Pt: 88…90 °C (11 mmHg) Density: 1.1575 g/cm³ Flash Pt: 84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000751 CAS Number: 14794-31-1 EINECS: 238-855-9 UN: 1760 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4350 to 1.4390 (20°C, 589 nm) |
GC | ≥94 % |
Titration RCOCl | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...