3-Methyl-1,2-cyclopentanedione 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Methyl-1,2-cyclopentanedione, Maple lactone, Methyl cyclopentenolone, 3-Methylcyclopentane-1,2-dione
17646-0100
17646-0500
TS17646-0100EA
51
USD
TS17646-0100
TS17646-0500
3-Methyl-1,2-cyclopentanedione 99%
3-Methyl-1,2-cyclopentanedione
Formula:
C₆H₈O₂ MW: 112.13 g/mol Melting Pt: 105…110 °C Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001417 CAS Number: 765-70-8 EINECS: 212-154-8 |
Specification Test Results
Appearance | White to yellow Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 104 to 108 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...