1,1,1-Trifluoroiodoethane 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17608-0100
17608-0500
TS17608-0100EA
67
USD
TS17608-0100
TS17608-0500
1,1,1-Trifluoroiodoethane 99%
1,1,1-Trifluoroiodoethane
Formula:
C₂H₂F₃I Boiling Pt: 53…55 °C (760 mmHg) Density: 2.13 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00001082 CAS Number: 353-83-3 EINECS: 206-541-0 |
Specification Test Results
Appearance | Clear colorless to pink Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4000 to 1.4020 (20°C, 589 nm) |
GC | ≥98.5 % |
Iron (Fe) | ≤0.02 % |
Heavy metals (as Pb) | ≤0.003 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...