Sulphobromophthalein sodium hydrate

Supplier: Thermo Scientific Chemicals
Danger

17340-0050 17340-0250
TS17340-0050EA 405 USD
TS17340-0050 TS17340-0250
Sulphobromophthalein sodium hydrate
Sulfobromophthalein sodium hydrate
Formula: C₂₀H₈Br₄Na₂O₁₀S₂·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD16556312
CAS Number: 123359-42-2

Order Now

Specification Test Results

Appearance White to off-white to grey Powder
Infrared spectrum Conforms
Water ≤8 % (K.F.)
HPLC ≥95 %
UV Lambda max 575 to 581 nm (c=10 mg/l, 0.1N NaOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR