Adipoyl dichloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Adipoyl chloride

17318-1000 17318-5000
TS17318-1000EA 223 USD
TS17318-1000 TS17318-5000
Adipoyl dichloride 98%
Adipoyl dichloride
Formula: C₆H₈Cl₂O₂
Boiling Pt: 105…107 °C (2 mmHg)
Density: 1.25 g/cm³
Flash Pt: 72 °C
MDL Number: MFCD00000759
CAS Number: 111-50-2
EINECS: 203-876-4
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear brown Liquid
Infrared spectrum Conforms
Titration RCOCl ≥97.5 %
Titration RCOOH ≤2 % (as Adipic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR