4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Amino-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Amino-1-oxy-2,2,6,6-tetramethylpiperidine

17273-0010 17273-0050 17273-2500
TS17273-0010EA 134 USD
TS17273-0010 TS17273-0050 TS17273-2500
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) 97% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO)
Formula: C₉H₁₉N₂O
MW: 171.26 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Appearance Red to red-brown Adhering crystals or needles or moist mass
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR