4-Biphenylcarbonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

17125-0100 17125-0500
TS17125-0100EA 61 USD
TS17125-0100 TS17125-0500
4-Biphenylcarbonyl chloride 98%
4-Biphenylcarbonyl chloride
Formula: C₁₃H₉ClO
Boiling Pt: 195…200 °C (5 mmHg)
Melting Pt: 110…114 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000692
CAS Number: 14002-51-8
EINECS: 237-804-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 114 °C
Titration Argentometric ≥97.5 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR