2-Cyclohepten-1-one 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17107-0010
17107-0050
TS17107-0010EA
78
USD
TS17107-0010
TS17107-0050
2-Cyclohepten-1-one 90%
2-Cyclohepten-1-one
Formula:
C₇H₁₀O Boiling Pt: 189 °C (760 mmHg) Density: 0.99 g/cm³ Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004157 CAS Number: 1121-66-0 |
Specification Test Results
Appearance | Colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥89.0 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...