2-Bromothiazole 98+%

Supplier: Thermo Scientific Chemicals
Warning

17091-0050 17091-0250
TS17091-0050EA 50 USD
TS17091-0050 TS17091-0250
2-Bromothiazole 98+%
2-Bromothiazole
Formula: C₃H₂BrNS
Boiling Pt: 70…71 °C (20 mmHg)
Density: 1.82 g/cm³
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005316
CAS Number: 3034-53-5
EINECS: 221-229-4

Order Now

Specification Test Results

Appearance Clear colorless to orange to light brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5910 to 1.5930 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR