Toluene-α-sulphonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

17049-0050 17049-0250
TS17049-0050EA 47 USD
TS17049-0050 TS17049-0250
Toluene-α-sulphonyl chloride 98%
Formula: C₇H₇ClO₂S
Boiling Pt: 120 °C (10 mmHg)
Melting Pt: 90…94 °C
MDL Number: MFCD00007455
CAS Number: 1939-99-7
EINECS: 217-717-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
Titration Argentometric ≥97.5 % (ex Cl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR