Phenoxyacetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

16994-0100 16994-0500 16994-2500
TS16994-0100EA 30 USD
TS16994-0100 TS16994-0500 TS16994-2500
Phenoxyacetyl chloride 98%
Phenoxyacetyl chloride
Formula: C₈H₇ClO₂
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 28…30 °C
Density: 1.23 g/cm³
Flash Pt: 108 °C
MDL Number: MFCD00000726
CAS Number: 701-99-5
EINECS: 211-862-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear light yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.5324 to 1.5344 (20°C, 589 nm)
Titration RCOCl ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR