Tributyl phosphate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: TBP, TBPA

16993-0010 16993-0025 16993-0100
TS16993-0010EA 79 USD
TS16993-0010 TS16993-0025 TS16993-0100
Tributyl phosphate 99+%
Tributyl phosphate
Formula: (CH₃(CH₂)₃O)₃PO
Boiling Pt: 289 °C (760 mmHg)
Melting Pt: –79 °C
Density: 0.979 g/cm³
Flash Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00009436
CAS Number: 126-73-8
EINECS: 204-800-2
Merck Index: 13,09692
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fb9a0e9a8476470c1c50ab98587c559fec16cd8d&vid=6bcd8ac2f1665c53ba2c7ace4f34ebb3d890b549&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4250 (20°C, 589 nm)
Water ≤0.2 %
GC ≥99.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR