Methyl-2-(triphenylphosphoranylidene)acetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane
16956-0250
16956-1000
16956-5000
TS16956-0250EA
110
USD
TS16956-0250
TS16956-1000
TS16956-5000
Methyl-2-(triphenylphosphoranylidene)acetate 98%
Methyl-2-(triphenylphosphoranylidene)acetate
Formula:
C₂₁H₁₉O₂P Melting Pt: 164…169 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008455 CAS Number: 2605-67-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 164 to 169 °C |
HPLC | ≤2 % triphenylphosphine + triphenylphosphine oxide |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...