Valeryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentanoyl chloride, Valeroyl chloride

16912-0010 16912-1000
TS16912-0010EA 372 USD
TS16912-0010 TS16912-1000
Valeryl chloride 98%
Valeryl chloride
Formula: C₅H₉ClO
MW: 120.58 g/mol
Boiling Pt: 125
Melting Pt: -110
Density: 0.99 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00000757
CAS Number: 638-29-9
EINECS: 211-330-1
UN: 2502
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4230 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR