4-tert-Butylbenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-tert-Butylbenzoylchloride

16910-0050 16910-0250 16910-1000
TS16910-0050EA 30 USD
TS16910-0050 TS16910-0250 TS16910-1000
4-tert-Butylbenzoyl chloride 98%
4-tert-Butylbenzoyl chloride
Formula: C₁₁H₁₃ClO
MW: 196.68 g/mol
Boiling Pt: 135 °C (20 mmHg)
Density: 1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000695
CAS Number: 1710-98-1
EINECS: 216-973-1
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5354 to 1.5374 (20°C, 589 nm)
Titration RCOCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR