1-Bromo-2-methylpropane 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Isobutyl bromide

16883-0025 16883-1000 16883-5000
TS16883-0025EA 460 USD
TS16883-0025 TS16883-1000 TS16883-5000
1-Bromo-2-methylpropane 98+%
1-Bromo-2-methylpropane
Formula: C₄H₉Br
MW: 137.02 g/mol
Boiling Pt: 90…92 °C (760 mmHg)
Melting Pt: –119 °C
Density: 1.26 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00000217
CAS Number: 78-77-3
EINECS: 201-141-2
UN: 2342
ADR: 3,II
Merck Index: 13,05152

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4370 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR