1-Bromooctane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Octyl bromide

16681-0025 16681-5000
TS16681-0025EA 323 USD
TS16681-0025 TS16681-5000
1-Bromooctane 99%
1-Bromooctane
Formula: BrH₂C(CH₂)₆CH₃
MW: 193.13 g/mol
Boiling Pt: 201 °C (760 mmHg)
Melting Pt: –55 °C
Density: 1.11 g/cm³
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00000276
CAS Number: 111-83-1
EINECS: 203-912-9
UN: 3082
ADR: 9,III
Merck Index: 12,06861

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4508 to 1.4528 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR