Fumaroyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Fumaryl chloride

16597-0250 16597-1000 16597-5000
TS16597-0250EA 51 USD
TS16597-0250 TS16597-1000 TS16597-5000
Fumaroyl chloride 95%
Fumaroyl chloride
Formula: C₄H₂Cl₂O₂
MW: 152.96 g/mol
Boiling Pt: 161…164 °C (760 mmHg)
Melting Pt: –2 °C
Density: 1.41 g/cm³
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000733
CAS Number: 627-63-4
EINECS: 211-005-4
UN: 1780
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5010 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR