Phenyl salicylate 99%

Supplier: Thermo Scientific Chemicals
Warning

16508-0010 16508-0025 16508-2500
TS16508-0010EA 171 USD
TS16508-0010 TS16508-0025 TS16508-2500
Phenyl salicylate 99%
Phenyl salicylate
Formula: C₁₃H₁₀O₃
Boiling Pt: 172…173 °C (12 mmHg)
Melting Pt: 40…44 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00002213
CAS Number: 118-55-8
EINECS: 204-259-2
Merck Index: 12,07464

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 40 to 44 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR