2-Iodopropane 98+% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Isopropyl iodide

16496-1000 16496-5000
TS16496-1000EA 81 USD
TS16496-1000 TS16496-5000
2-Iodopropane 98+% stabilized
2-Iodopropane
Formula: C₃H₇I
MW: 169.99 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –90 °C
Density: 1.7 g/cm³
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00001071
CAS Number: 75-30-9
EINECS: 200-859-3
UN: 2392
ADR: 3,III
Merck Index: 13,05234

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4972 to 1.4992 (20°C, 589 nm)
GC ≥98.0 %
Stabilizer copper
Elemental Analysis 6.15 to 6.94 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR