Thiobenzamide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Benzothioamide
16473-0100
16473-0500
TS16473-0100EA
30
USD
TS16473-0100
TS16473-0500
Thiobenzamide 95%
Thiobenzamide
Formula:
C₇H₇NS MW: 137.21 g/mol Boiling Pt: 245 °C (1013 hPa) Melting Pt: 114…118 °C |
MDL Number:
MFCD00008060 CAS Number: 2227-79-4 EINECS: 218-765-6 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Yellow-green Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 114 to 118 °C |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...