Heptanoyl chloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Heptanoylchloride
16426-0250
16426-1000
16426-5000
TS16426-0250EA
39
USD
TS16426-0250
TS16426-1000
TS16426-5000
Heptanoyl chloride 99%
Heptanoyl chloride
Formula:
C₇H₁₃ClO MW: 148.63 g/mol Boiling Pt: 173 Melting Pt: -83.8 Density: 0.96 g/cm³ Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000763 CAS Number: 2528-61-2 EINECS: 219-775-3 UN: 2927 ADR: 6.1 II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4290 to 1.4320 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...