Quinoxaline-2-carbonyl chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Quinoxaloyl chloride

16376-0010 16376-2500
TS16376-0010EA 191 USD
TS16376-0010 TS16376-2500
Quinoxaline-2-carbonyl chloride 95%
Quinoxaline-2-carbonyl chloride
Formula: C₉H₅ClN₂O
MW: 192.61 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006725
CAS Number: 54745-92-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
GC ≥94.0 %
Insoluble matter ≤0.05 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR