1-Bromodecane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Decyl bromide

16360-0010 16360-5000
TS16360-0010EA 103 USD
TS16360-0010 TS16360-5000
1-Bromodecane 98%
1-Bromodecane
Formula: BrH₂C(CH₂)₈CH₃
MW: 221.18 g/mol
Boiling Pt: 238
Melting Pt: -30
Density: 1.06 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00000221
CAS Number: 112-29-8
EINECS: 203-955-3
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4550 to 1.4570 (20°C, 589 nm)
GC 97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR