3-Methyl-2-cyclopenten-1-one 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone

16359-0100 16359-0500 16359-2500
TS16359-0100EA 87 USD
TS16359-0100 TS16359-0500 TS16359-2500
3-Methyl-2-cyclopenten-1-one 98% stabilized
3-Methyl-2-cyclopenten-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 74 °C (15 mmHg)
Melting Pt: 3…5 °C
Density: 0.98 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001403
CAS Number: 2758-18-1
EINECS: 220-421-5

Order Now

Specification Test Results

Appearance Clear light yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4900 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % hydroquinone
UV at 240 nm A: ≤0.26
Wavelength at 1 A.U. ≤215 nm
Residue after evaporation ≤2 ppm
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0006 meq/g
Peroxides ≤0.0075 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR