2,4-Dichlorotoluene 99%

Supplier: Thermo Scientific Chemicals
Warning

16328-0010 16328-2500
TS16328-0010EA 142 USD
TS16328-0010 TS16328-2500
2,4-Dichlorotoluene 99%
2,4-Dichlorotoluene
Formula: C₇H₆Cl₂
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: –14 °C
Density: 1.24 g/cm³
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000583
CAS Number: 95-73-8
EINECS: 202-445-8
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5470 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR