2,4-Dichloropyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

16265-0050 16265-0250 16265-1000
TS16265-0050EA 44 USD
TS16265-0050 TS16265-0250 TS16265-1000
2,4-Dichloropyrimidine 97%
2,4-Dichloropyrimidine
Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 101 °C (23 mmHg)
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006061
CAS Number: 3934-20-1
EINECS: 223-508-6

Order Now

Specification Test Results

Appearance White to yellow to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 57 to 62 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR