4,4'-Dipyridyl disulphide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,4-Dipyridyl Disulphide, 4,4'-Dithiodipyridine

16224-0050 16224-0010 16224-0250
TS16224-0050EA 166 USD
TS16224-0050 TS16224-0010 TS16224-0250
4,4'-Dipyridyl disulphide 98%
4,4'-Dipyridyl disulphide
Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Boiling Pt: 348 °C (1013 hPa)
Melting Pt: 75…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006423
CAS Number: 2645-22-9
EINECS: 220-158-6
UN: 3335

Order Now

Specification Test Results

Appearance White to orange Crystalline powder
Infrared spectrum Conforms
Melting point 75 to 79 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR