2,2'-Dipyridyl disulphide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Di(pyridin-2-yl)disulphane, 2,2'-Dithiodipyridine

16223-0050 16223-0250 16223-1000
TS16223-0050EA 108 USD
TS16223-0050 TS16223-0250 TS16223-1000
2,2'-Dipyridyl disulphide 98%
2,2'-Dipyridyl disulphide
Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Melting Pt: 56…60 °C
Flash Pt: 210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 55 to 60 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR