2,6-Pyridinedicarbonyl dichloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,6-Pyridinedicarboxylicacidchloride, 2,6-Pyridinedicarboxylic acid chloride

16213-0050 16213-0250 16213-1000
TS16213-0050EA 68 USD
TS16213-0050 TS16213-0250 TS16213-1000
2,6-Pyridinedicarbonyl dichloride 97%
2,6-Pyridinedicarbonyl dichloride
Formula: C₇H₃Cl₂NO₂
MW: 204.006 g/mol
Boiling Pt: 284 °C (760 mmHg)
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00006289
CAS Number: 3739-94-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 57 to 61 °C
Titration RCOCl ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR