Atrazine

Supplier: Thermo Scientific Chemicals
Warning

16168-0050 16168-0250
TS16168-0050EA 99 USD
TS16168-0050 TS16168-0250
Atrazine
Atrazine
Formula: C₈H₁₄ClN₅
Melting Pt: 175 °C
Storage Temperature: Ambient
MDL Number: MFCD00041810
CAS Number: 1912-24-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Infrared spectrum Conforms
Melting point 173 to 180 °C
Water ≤1 %
HPLC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR