Phenyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

16097-5000 16097-0025 16097-1000
TS16097-5000EA 224 USD
TS16097-5000 TS16097-0025 TS16097-1000
Phenyl isothiocyanate 98%
Phenyl isothiocyanate
Formula: C₇H₅NS
Boiling Pt: 221 °C (760 mmHg)
Melting Pt: –21 °C
Density: 1.1288 g/cm³
Flash Pt: 87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004798
CAS Number: 103-72-0
EINECS: 203-138-1
UN: 2927
ADR: 6.1,II
Merck Index: 12,07450

Order Now

Specification Test Results

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.6490 to 1.6510 (20°C, 589 nm)
Water ≤0.1 %
GC ≥97.5 %
Specific gravity (20°C) 1.13 to 1.136


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR