Methyl-2,2,2-trichloroacetimidate 98%

Supplier: Thermo Scientific Chemicals
Warning

16093-0250 16093-1000
TS16093-0250EA 121 USD
TS16093-0250 TS16093-1000
Methyl-2,2,2-trichloroacetimidate 98%
Methyl-2,2,2-trichloroacetimidate
Formula: C₃H₄Cl₃NO
Boiling Pt: 148…149 °C (760 mmHg)
Density: 1.42 g/cm³
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001759
CAS Number: 2533-69-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4800 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR