Isobutyryl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

16066-0010 16066-0100 16066-2500
TS16066-0010EA 188 USD
TS16066-0010 TS16066-0100 TS16066-2500
Isobutyryl chloride 98%
Isobutyryl chloride
Formula: C₄H₇ClO
MW: 106.55 g/mol
Melting Pt: –90 °C
Density: 1.017 g/cm³
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00000717
CAS Number: 79-30-1
EINECS: 201-194-1
UN: 2395
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4060 to 1.4080 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR